missing translation for 'onlineSavingsMsg'
Learn More

2-Azahypoxanthine Sodium Salt, TRC

Product Code. 30137801
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137801 1 g 1g
30006895 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137801 Supplier Toronto Research Chemicals Supplier No. TRCA8001001G
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797817-35-6
Molecular Formula C4H2N5NaO
Synonym 3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one Sodium Salt∣1,5-Dihydro-4H-Imidazo[4,5-d]- 1,2,3-triazin-4-one∣3,7-Dihydro-4H-Imidazo[4,5-d]-v-triazin-4-one Sodium Salt∣NSC 22419 Sodium Salt∣AHX sodium salt

Specifications

Format Neat
Molecular Weight (g/mol) 159.08
Shipping Condition Room Temperature
Quantity 1 g
Chemical Name or Material 2-Azahypoxanthine Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.