missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylthiophene, 98%

Product Code. 11499902
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499902 10 g 10g
11319679 5 g 5g
11409912 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499902 Supplier Thermo Scientific Alfa Aesar Supplier No. A12036.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 135-00-2 | C11H8OS | 188.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-00-2
Molecular Formula C11H8OS
Molecular Weight (g/mol) 188.244
MDL Number MFCD00022495
InChI Key DWYFUJJWTRPARQ-UHFFFAOYSA-N
Synonym 2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene
PubChem CID 67262
IUPAC Name phenyl(thiophen-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CS2

Specifications

Melting Point 55°C to 57°C
Boiling Point 124°C to 126°C (0.5 mmHg)
Quantity 10 g
Beilstein 3929
Formula Weight 188.25
Percent Purity 98%
Chemical Name or Material 2-Benzoylthiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.