missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-L-proline hydrochloride, 95%

Product Code. 15448607
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448607 250 mg 250mg
15438607 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448607 Supplier Thermo Scientific Alfa Aesar Supplier No. H52080.MD
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 86116-84-9 | C12H15NO2 | 205.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86116-84-9
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD16660381
InChI Key LIHLFIASPODZOB-GFCCVEGCSA-N
Synonym 2-benzylproline, 2-benzyl-l-proline, r-2-benzylpyrrolidine-2-carboxylic acid, l-proline, 2-phenylmethyl, r-2-benzyl-pyrrolidine-2-carboxylic acid, r-alpha-benzyl-l-proline-hcl, r-a-benzyl-prolinecl, 2r-2-benzylpyrrolidine-2-carboxylic acid, r-a-benzyl proline?hcl, r-alpha-benzylproline hcl
PubChem CID 2761822
IUPAC Name (2R)-2-benzylpyrrolidine-2-carboxylic acid
SMILES C1CC(NC1)(CC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 270°C to 272°C
Quantity 250 mg
Formula Weight 241.72
Optical Rotation −33° (c=1 in 1N HCl)
Percent Purity 95%
Chemical Name or Material 2-Benzyl-L-proline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.