missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzyloxy)phenol, TRC

Product Code. 30347392
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347392 10 g 10g
30338479 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30347392 Supplier Toronto Research Chemicals Supplier No. TRCB28665510G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Benzyloxy)phenol
CAS 6272-38-4
Molecular Formula C13H12O2
Quantity 10 g
Synonym Benzyl o-Hydroxyphenyl Ether, NSC 37426, NSC 72669, o-(Benzyloxy)phenol
InChI Formula InChI=1S/C13H12O2/c14-12-8-4-5-9-13(12)15-10-11-6-2-1-3-7-11/h1-9,14H,10H2
SMILES Oc1ccccc1OCc2ccccc2
IUPAC Name 2-phenylmethoxyphenol
Molecular Weight (g/mol) 200.23
Formula Weight 200.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.