missing translation for 'onlineSavingsMsg'
Learn More

2-[Bis(methylthio)methylene]malononitrile, 97%, Thermo Scientific Chemicals

Product Code. 15430398
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
15430398 25 g 25g
15420398 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15430398

Brand: Thermo Scientific Alfa Aesar H53503.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-[Bis(methylthio)methylene]malononitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-[Bis(methylthio)methylene]malononitrile is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5147-80-8
Molecular Formula C6H6N2S2
Molecular Weight (g/mol) 170.248
MDL Number MFCD00052730
InChI Key FICQFRCPSFCFBY-UHFFFAOYSA-N
Synonym 2-bis methylthio methylene malononitrile, bis methylthio methylene malononitrile, 2-di methylthio methylidene malononitrile, propanedinitrile, bis methylthio methylene, 2-bis methylsulfanyl methylidene propanedinitrile, bis methylthio methylene propanedinitrile, 3,3-bis methylthio-2-cyanoacrylonitrile, bis methylthio methylidene propanedinitrile
PubChem CID 99229
IUPAC Name 2-[bis(methylsulfanyl)methylidene]propanedinitrile
SMILES CSC(=C(C#N)C#N)SC

Specifications

Melting Point 79°C to 82°C
Linear Formula (NC)2C=C(SCH3)2
Quantity 25 g
UN Number UN3439
Formula Weight 170.25
Percent Purity 97%
Chemical Name or Material 2-[Bis(methylthio)methylene]malononitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.