missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)-5-(pentafluoroethyl)pyridine, 96%, Thermo Scientific™

Product Code. 15450097
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450097 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15450097 Supplier Thermo Scientific Alfa Aesar Supplier No. H66291.MC
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1580464-68-1
Molecular Formula C12H13F5N2O2
Quantity 100 mg
InChI Key ZASNKMWFNLRBGM-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC1=NC=C(C=C1)C(C(F)(F)F)(F)F
IUPAC Name tert-butyl N-[5-(1,1,2,2,2-pentafluoroethyl)pyridin-2-yl]carbamate
Molecular Weight (g/mol) 312.24
PubChem CID 99728550
Formula Weight 312.24
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-(Boc-amino)-5-(pentafluoroethyl)pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.