missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(1,3-thiazol-2-yl)ethanone, ≥97%, Thermo Scientific™

Product Code. 10689564
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10689564 250 mg 250mg
10699564 1 g 1g
10567054 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10689564 Supplier Thermo Scientific Maybridge Supplier No. CC09505CB
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 3292-77-1
Molecular Formula C5H4BrNOS
Molecular Weight (g/mol) 206.06
MDL Number MFCD06411540
InChI Key AQRFTRDAOYSMEA-UHFFFAOYSA-N
Synonym 2-bromo-1-thiazol-2-yl ethanone, 2-bromo-1-1,3-thiazol-2-yl ethanone, 2-bromoacetyl-1,3-thiazole, 2-bromo-1-thiazol-2-yl-ethanone, 2-bromo-1-1,3-thiazol-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thiazolyl, 2-bromo-1-1,3-thiazol-2-yl-1-ethanone, 1-bromo-1,3-thiazole-2-yl ethane-2-one, 2-bromoacetylthiazole, 2-bromoacetyl thiazole
PubChem CID 2795212
IUPAC Name 2-bromo-1-(1,3-thiazol-2-yl)ethanone
SMILES BrCC(=O)C1=NC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 206.06
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(1, 3-thiazol-2-yl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.