missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(1,3-thiazol-2-yl)ethanone, TRC

Product Code. 30338793
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338793 1 g 1g
30329798 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30338793 Supplier Toronto Research Chemicals Supplier No. TRCB6884081G
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromo-1-(1,3-thiazol-2-yl)ethanone
CAS 3292-77-1
Molecular Formula C5H4BrNOS
Quantity 1 g
InChI Formula InChI=1S/C5H4BrNOS/c6-3-4(8)5-7-1-2-9-5/h1-2H,3H2
SMILES BrCC(=O)c1nccs1
IUPAC Name 2-bromo-1-(1,3-thiazol-2-yl)ethanone
Molecular Weight (g/mol) 206.06
Formula Weight 204.9197
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.