missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2,3-dihydro-1,4-benzodioxin-5-yl)-1-ethanone, 97%, Thermo Scientific™

Product Code. 10374562
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10374562 1 g Amber Glass Bottle 1g
10692433 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10374562 Supplier Thermo Scientific Maybridge Supplier No. CC01405DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 19815-97-5
Molecular Formula C10H9BrO3
Molecular Weight (g/mol) 257.08
MDL Number MFCD04972617
InChI Key BSROYFIAEPSLCT-UHFFFAOYSA-N
Synonym 2-bromo-1-2,3-dihydrobenzo b 1,4 dioxin-5-yl ethanone, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-5-yl-1-ethanone, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-5-yl ethanone, 5-bromoacetyl-1,4-benzodioxane, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-5-yl ethan-1-one, 5-bromoacetyl-2,3-dihydro-1,4-benzodioxine, 2-bromanyl-1-2,3-dihydro-1,4-benzodioxin-5-yl ethanone, 1-2h,3h-benzo 2,3-e 1,4-dioxin-5-yl-2-bromoethan-1-one, ethanone, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-5-yl
PubChem CID 2795035
IUPAC Name 2-bromo-1-(2,3-dihydro-1,4-benzodioxin-5-yl)ethanone
SMILES BrCC(=O)C1=C2OCCOC2=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 257.08
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(2, 3-dihydro-1, 4-benzodioxin-5-yl)-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.