missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl)-1-ethanone, 97%, Thermo Scientific™

Product Code. 10492033
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10492033 250 mg Amber Glass Bottle 250mg
10644553 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10492033 Supplier Thermo Scientific Maybridge Supplier No. MO00935CB
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 54001-36-4
Molecular Formula C12H9BrClNOS
Molecular Weight (g/mol) 330.62
MDL Number MFCD04115414
InChI Key MDZFMZALZSZQSM-UHFFFAOYSA-N
Synonym 2-bromo-1-2-4-chlorophenyl-4-methyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-2-4-chlorophenyl-4-methyl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-4'-chlorophenyl thiazole, 2-bromo-1-2-4-chlorophenyl-4-methylthiazol-5-yl ethanone, 2-bromo-1-2-4-chlorophenyl-4-methyl-5-thiazolyl ethanone, 2-bromo-1-2-4-chlorophenyl-4-methyl-thiazol-5-yl ethanone, 2-bromanyl-1-2-4-chlorophenyl-4-methyl-1,3-thiazol-5-yl ethanone, 2-bromo-1-2-4-chlorophenyl-4-methyl-1,3-thiazol-5-yl ethan-1-one
PubChem CID 2794647
IUPAC Name 2-bromo-1-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]ethanone
SMILES CC1=C(SC(=N1)C1=CC=C(Cl)C=C1)C(=O)CBr

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 330.63
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-[2-(4-chlorophenyl)-4-methyl-1, 3-thiazol-5-yl]-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.