missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide, Thermo Scientific™

Product Code. 10314762
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10314762 1 g Amber Glass Bottle 1g
10471304 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10314762 Supplier Thermo Scientific Maybridge Supplier No. CC04005DA
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 17570-98-8
Molecular Formula C7H7Br2NO
Molecular Weight (g/mol) 280.947
InChI Key BYKVUGZUYJUSKD-UHFFFAOYSA-N
Synonym 2-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-2-yl ethanone hydrobromide, 2-bromo-1-2-pyridinyl-1-ethanone hydrobromide, 2-2-bromoacetyl pyridine hydrobromide, 2-bromoacetylpyridine hydrobromide, 2-bromo-1-pyridin-2-ylethanone hydrobromide, 2-bromo-1-2-pyridinyl ethanone hydrobromide, 2-bromo-1-pyridin-2-yl ethan-1-one hydrobromide, zlchem 1071, pubchem21374
PubChem CID 2776231
IUPAC Name 2-bromo-1-pyridin-2-ylethanone;hydrobromide
SMILES C1=CC=NC(=C1)C(=O)CBr.Br

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 280.95
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.