missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[3-(3,4-dichlorophenyl)isoxazol-5-yl]ethan-1-one, Tech., Thermo Scientific™

Product Code. 10069601
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10069601 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10069601 Supplier Thermo Scientific Maybridge Supplier No. SPB04954DA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 175277-38-0
Packaging Amber Glass Bottle
Molecular Formula C11H6BrCl2NO2
MDL Number MFCD00173899
Quantity 1 g
Synonym 5-bromoacetyl-3-3,4-dichlorophenyl isoxazole, 2-bromo-1-3-3,4-dichlorophenyl isoxazol-5-yl ethan-1-one, 2-bromo-1-3-3,4-dichlorophenyl-isoxazol-5-yl ethanone, 2-bromo-1-3-3,4-dichlorophenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-3,4-dichlorophenyl isoxazol-5-yl ethanone, pubchem8719, buttpark 43\57-73
InChI Key CBDHLEMYIQJYII-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C2=NOC(=C2)C(=O)CBr)Cl)Cl
IUPAC Name 2-bromo-1-[3-(3,4-dichlorophenyl)-1,2-oxazol-5-yl]ethanone
Molecular Weight (g/mol) 334.978
PubChem CID 2799583
Formula Weight 334.98
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-1-[3-(3, 4-dichlorophenyl)isoxazol-5-yl]ethan-1-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.