missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[3,5-di(trifluoromethyl)phenyl]ethan-1-one, 95%, Thermo Scientific™

Product Code. 10693033
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10693033 1 g 1g
10334112 10 g 10g
10672505 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10693033 Supplier Thermo Scientific Maybridge Supplier No. SPB07614DA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 131805-94-2
Molecular Formula C10H5BrF6O
Molecular Weight (g/mol) 335.043
MDL Number MFCD00792434
InChI Key ZEKBFXJTIAEUOF-UHFFFAOYSA-N
Synonym 1-3,5-bis trifluoromethyl phenyl-2-bromoethanone, 3',5'-bis trifluoromethyl-2-bromoacetophenone, 1-3,5-bis trifluoromethyl phenyl-2-bromoethan-1-one, 3,5-bistrifluoromethyl-phenacyl bromide, 3,5-bis trifluoromethyl phenacyl bromide, ethanone,1-3,5-bis trifluoromethyl phenyl-2-bromo, 3,5-bis trifluoromethyl phenacylbromide, a-bromo-3',5'-bis trifluoromethyl acetophenone, 2-bromo-1-3,5-bis trifluoromethyl phenyl ethanone
PubChem CID 2736170
IUPAC Name 1-[3,5-bis(trifluoromethyl)phenyl]-2-bromoethanone
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)CBr

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula (CF3)2C6H3C(O)CH2Br
Quantity 1 g
Formula Weight 335.04
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-[3, 5-di(trifluoromethyl)phenyl]ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.