missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one, Tech., Thermo Scientific™

Product Code. 10793993
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging missing translation for 'unitSize'
10793993 10 g Amber Glass Bottle 10g
10222492 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10793993 missing translation for 'mfr' Thermo Scientific Maybridge missing translation for 'supplierNo' BTB09098EA
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175135-93-0
Molecular Formula C10H10BrClO
Molecular Weight (g/mol) 261.543
MDL Number MFCD00067862
InChI Key RGMFWCVOMGKIJM-UHFFFAOYSA-N
Synonym 2-bromo-1-3-chloro-4-methylphenyl propan-1-one, 2-bromo-3'-chloro-4'-methylpropiophenone, 1-propanone,2-bromo-1-3-chloro-4-methylphenyl, 2-bromo-3'-chloro-4'methyl propiophenone, 2-bromo-1-3-chloro-4-methylphenyl-1-propanone, 2-bromanyl-1-3-chloranyl-4-methyl-phenyl propan-1-one
PubChem CID 2774950
IUPAC Name 2-bromo-1-(3-chloro-4-methylphenyl)propan-1-one
SMILES CC1=C(C=C(C=C1)C(=O)C(C)Br)Cl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 261.55
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.