missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-chlorophenyl)-2-(4-methylphenyl)ethan-1-one, ≥95%, Thermo Scientific™

Product Code. 10190872
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10190872 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10190872 Supplier Thermo Scientific Maybridge Supplier No. DP00551DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 119267-79-7
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C15H12BrClO
Linear Formula CH3C6H4CHBrC(O)C6H4Cl
MDL Number MFCD00052202
Quantity 1 g
Synonym 2-bromo-1-4-chlorophenyl-2-4-methylphenyl ethan-1-one, 2-bromo-1-4-chlorophenyl-2-4-methylphenyl ethanone, ethanone,2-bromo-1-4-chlorophenyl-2-4-methylphenyl, acmc-1c1ge, alpha-bromo-4-chloro-4'-methyldeoxybenzoin, alpha-bromo-4-chloro-4/'-methyldeoxybenzoin, 2-bromo-1-4-chlorophenyl-2-4-tolyl ethanone, 2-bromo-1-4-chlorophenyl-2-p-tolyl ethanone
InChI Key DGZRBDCHGUEXDT-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)C(C(=O)C2=CC=C(C=C2)Cl)Br
IUPAC Name 2-bromo-1-(4-chlorophenyl)-2-(4-methylphenyl)ethanone
Molecular Weight (g/mol) 323.614
PubChem CID 2777296
Formula Weight 323.62
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-chlorophenyl)-2-(4-methylphenyl)ethan-1-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.