missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-chlorophenyl)-2-phenylethan-1-one, ≥95%, Thermo Scientific™

Product Code. 10394372
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10394372 1 g Amber Glass Bottle 1g
10334332 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10394372 Supplier Thermo Scientific Maybridge Supplier No. DP00542DA
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1889-78-7
Molecular Formula C14H10BrClO
Molecular Weight (g/mol) 309.59
MDL Number MFCD00052201
InChI Key OBEFSOTWERFWSZ-UHFFFAOYNA-N
Synonym 2-bromo-1-4-chlorophenyl-2-phenylethan-1-one, 2-bromo-1-4-chlorophenyl-2-phenylethanone, 1-4-chlorophenyl-2-bromo-2-phenylethanone, 2-bromo-1-4-chlorophenyl-2-phenyl-ethanone, alpha-bromo-alpha-phenyl-4'-chloroacetophenone, ethanone,2-bromo-1-4-chlorophenyl-2-phenyl
PubChem CID 2777295
SMILES ClC1=CC=C(C=C1)C(=O)C(Br)C1=CC=CC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula C6H5CHBrC(O)C6H4Cl
Quantity 1 g
IUPAC Name 2-bromo-1-(4-chlorophenyl)-2-phenylethanone
Formula Weight 309.59
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-chlorophenyl)-2-phenylethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.