missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-isobutylphenyl)propan-1-one, 97%, Thermo Scientific™

Product Code. 10682263
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10682263 10 g Amber Glass Bottle 10g
10446672 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10682263 Supplier Thermo Scientific Maybridge Supplier No. RH01005EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 80336-64-7
Molecular Formula C13H17BrO
Molecular Weight (g/mol) 269.18
MDL Number MFCD00831700
InChI Key SVGMFVCPJFHTRE-UHFFFAOYNA-N
Synonym 2-bromo-1-4-isobutylphenyl propan-1-one, 2-bromo-1-4-2-methylpropyl phenyl propan-1-one, 1-4-isobutylphenyl-2-bromo-1-propanone, 2-bromo-4'-2-methylprop-1-yl propiophenone, 2-bromo-1-4-2-methylpropyl phenyl-1-propanone, 2-bromanyl-1-4-2-methylpropyl phenyl propan-1-one, 1-propanone,2-bromo-1-4-1,1-dimethylethyl phenyl
PubChem CID 2735576
IUPAC Name 2-bromo-1-[4-(2-methylpropyl)phenyl]propan-1-one
SMILES CC(C)CC1=CC=C(C=C1)C(=O)C(C)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3CH(Br)C(O)C6H4CH2CH(CH3)2
Quantity 10 g
Formula Weight 269.18
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(4-isobutylphenyl)propan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.