missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1,3-thiazol-5-yl]-1-ethanone, ≥90%, Thermo Scientific™

Product Code. 10173384
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10173384 5 g 5g
10612443 250 mg 250mg
10622443 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10173384 Supplier Thermo Scientific Maybridge Supplier No. CC19805DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 423768-43-8
Molecular Formula C10H8BrN3OS
Molecular Weight (g/mol) 298.158
MDL Number MFCD03407321
InChI Key PQWKERRFBZJRSD-UHFFFAOYSA-N
Synonym 2-bromo-1-4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-2-pyrazinyl-5-thiazolyl, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 2-bromo-1 4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethan-1-one
PubChem CID 2776507
IUPAC Name 2-bromo-1-(4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)C2=NC=CN=C2)C(=O)CBr

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 5 g
Formula Weight 298.16
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1, 3-thiazol-5-yl]-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.