missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)-1-ethanone, 95%, Thermo Scientific™

Product Code. 10337672
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10337672 1 g 1g
10402453 250 mg 250mg
10596595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10337672 Supplier Thermo Scientific Maybridge Supplier No. CC29205DA
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7520-95-8
Molecular Formula C12H10BrNOS
Molecular Weight (g/mol) 296.182
MDL Number MFCD04071443
InChI Key BOMSILGSXFNLJX-UHFFFAOYSA-N
Synonym 2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-phenyl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-phenylthiazole, 2-bromo-1-4-methyl-2-phenylthiazol-5-yl ethanone, 2-bromo-1-4-methyl-2-phenyl 1,3-thiazol-5-yl ethan-1-one
PubChem CID 2795492
IUPAC Name 2-bromo-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)C2=CC=CC=C2)C(=O)CBr

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 296.19
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(4-methyl-2-phenyl-1, 3-thiazol-5-yl)-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.