missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-methylphenyl)ethan-1-one, 97%, Maybridge™

Product Code. 10386474
Click to view available options
Quantity:
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10386474

Brand: Thermo Scientific Maybridge CD00282EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 619-41-0
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
InChI Key KRVGXFREOJHJAX-UHFFFAOYSA-N
Synonym 2-bromo-4'-methylacetophenone, 2-bromo-1-p-tolyl ethanone, 4-methylphenacyl bromide, 2-bromo-1-4-methylphenyl ethanone, 2-bromo-1-p-tolylethanone, 2-bromo-4-methylacetophenone, 2-bromo-1-4-methylphenyl ethan-1-one, acetophenone, 2-bromo-4'-methyl, ethanone, 2-bromo-1-4-methylphenyl, p-methylphenacyl bromide
PubChem CID 69272
IUPAC Name 2-bromo-1-(4-methylphenyl)ethanone
SMILES CC1=CC=C(C=C1)C(=O)CBr

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3C6H4COCH2Br
Quantity 10 g
Formula Weight 213.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(4-methylphenyl)ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.