missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-morpholinophenyl)-1-ethanone, 97%, Thermo Scientific™

Product Code. 10792473
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10792473 1 g 1g
10394562 250 mg 250mg
10275244 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10792473 Supplier Thermo Scientific Maybridge Supplier No. CC17405DA
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 210832-85-2
Molecular Formula C12H14BrNO2
Molecular Weight (g/mol) 284.15
MDL Number MFCD03783555
InChI Key OUGMZFJPRSTGMJ-UHFFFAOYSA-N
Synonym 2-bromo-1-4-morpholinophenyl-1-ethanone, 2-bromo-1-4-morpholinophenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethanone, 2-bromo-1-4-4-morpholinyl phenyl ethanone, 2-bromo-1-4-morpholin-4-yl-phenyl ethanone, 2-bromo-1-4-morpholin-4-yl phenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethan-1-one, pubchem23338, acmc-1cqxm, alpha-bromo-4'-morpholinoacetophenone
PubChem CID 2795357
SMILES BrCC(=O)C1=CC=C(C=C1)N1CCOCC1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2-bromo-1-[4-(morpholin-4-yl)phenyl]ethan-1-one
Formula Weight 284.15
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-morpholinophenyl)-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.