missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-pentylphenyl)ethan-1-one, 95%, Thermo Scientific™

Product Code. 10190622
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10190622 10 g Amber Glass Bottle 10g
10180622 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10190622 Supplier Thermo Scientific Maybridge Supplier No. BTB12841EA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 64328-68-3
Molecular Formula C13H17BrO
Molecular Weight (g/mol) 269.182
MDL Number MFCD00218837
InChI Key AVYNDJWQMUOSJZ-UHFFFAOYSA-N
Synonym 2-bromo-1-4-pentylphenyl ethan-1-one, 2-bromo-1-4-pentylphenyl ethanone, 2-bromo-4'-pent-1-yl acetophenone, ethanone,2-bromo-1-4-pentylphenyl, 2-bromo-1-4-pent-1-yl phenyl ethan-1-one
PubChem CID 2775670
IUPAC Name 2-bromo-1-(4-pentylphenyl)ethanone
SMILES CCCCCC1=CC=C(C=C1)C(=O)CBr

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)4C6H4C(O)CH2Br
Quantity 10 g
Formula Weight 269.18
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-pentylphenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.