missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone, ≥95%, Thermo Scientific™

Product Code. 10742083
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10742083 250 mg 250mg
10150772 1 g 1g
10220415 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10742083 Supplier Thermo Scientific Maybridge Supplier No. CC15505CB
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 137577-00-5
Molecular Formula C12H11BrN2O
Molecular Weight (g/mol) 279.137
MDL Number MFCD02681921
InChI Key VYGXRQSIPNGJNK-UHFFFAOYSA-N
Synonym 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl-1-ethanone, 2-bromo-1-5-methyl-1-phenylpyrazol-4-yl ethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl ethan-1-one, ethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl, acmc-1c040, 1-phenyl-4-2-bromoacetyl-5-methylpyrazole, 4-bromoacetyl-5-methyl-1-phenyl-1h-pyrazole, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl ethanone, 1-1-phenyl-5-methyl-1h-pyrazole-4-yl-2-bromoethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl-ethanone
PubChem CID 2776439
IUPAC Name 2-bromo-1-(5-methyl-1-phenylpyrazol-4-yl)ethanone
SMILES CC1=C(C=NN1C2=CC=CC=C2)C(=O)CBr

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 279.14
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.