missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-butene, 97%

Product Code. 11339246
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11339246 1 g 1g
11410561 5 g 5g
11420561 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339246 Supplier Thermo Scientific Alfa Aesar Supplier No. B20067.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 23074-36-4 | C4H7Br | 135.004 g/mol

2-Bromo-1-butene is used to produce 2-Ethyl-acrylic acid. It acts as a photolytic precursor for 1-buten-2-yl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1-butene is used to produce 2-Ethyl-acrylic acid. It acts as a photolytic precursor for 1-buten-2-yl.

Solubility
Soluble in ethanol, acetone and benzene. Not miscible in water.

Notes
Store away from air. Store under dry inert gas. Incompatible with oxidizing agents and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23074-36-4
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.004
MDL Number MFCD00039178
InChI Key HQMXRIGBXOFKIU-UHFFFAOYSA-N
PubChem CID 89990
IUPAC Name 2-bromobut-1-ene
SMILES CCC(=C)Br

Specifications

Refractive Index 1.4525
Quantity 1 g
UN Number UN1993
Synonym 2-bromo-1-butene
Solubility Information Soluble in ethanol,acetone and benzene. Not miscible in water.
Formula Weight 135
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-butene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.