missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-methyl-1H-imidazole, 95%, ACROS Organics™

Product Code. 10555670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10555670 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10555670 Supplier Thermo Scientific Acros Supplier No. 435430050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16681-59-7
CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Pink
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C4H5BrN2
MDL Number MFCD02179525
Quantity 5 g
Synonym 2-bromo-1-methyl-1h-imidazole, 1-methyl-2-bromoimidazole, 2-bromo-n-methylimidazole, 1h-imidazole, 2-bromo-1-methyl, zlchem 620, pubchem7594, n-methyl-2-bromoimidazole, acmc-209dv3, 2-bromanyl-1-methyl-imidazole, 1-methyl-2-bromo-1h-imidazole
InChI Key BANOTGHIHYMTDL-UHFFFAOYSA-N
SMILES CN1C=CN=C1Br
IUPAC Name 2-bromo-1-methylimidazole
Molecular Weight (g/mol) 161.002
PubChem CID 2773262
Formula Weight 161
Percent Purity ≥94.0%
Physical Form Liquid
Chemical Name or Material 2-Bromo-1-methyl-1H-imidazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-1-methyl-1H-imidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.