missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-methylbenzimidazole, 97%

Product Code. 15476538
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476538 250 mg 250mg
15456538 1 g 1g
15466538 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15476538 Supplier Thermo Scientific Alfa Aesar Supplier No. H60075.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 49572-60-3 | C8H7BrN2 | 211.06 g/mol

It is a synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a synthetic building block.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49572-60-3
Molecular Formula C8H7BrN2
Molecular Weight (g/mol) 211.06
MDL Number MFCD00839528
InChI Key PROLKELCEPYEQT-UHFFFAOYSA-N
Synonym 2-bromo-1-methyl-1h-benzo d imidazole, 2-bromo-1-methyl-1h-benzoimidazole, 1h-benzimidazole, 2-bromo-1-methyl, 2-bromo-1-methyl-1h-benzimidazole, 2-bromo-1-methyl-1,3-benzodiazole, 1-methyl-2-bromo-1h-benzimidazole, 2-bromo-1-methyl-1h-1,3-benzodiazole
PubChem CID 797082
SMILES CN1C(Br)=NC2=CC=CC=C12

Specifications

Quantity 250 mg
IUPAC Name 2-bromo-1-methylbenzimidazole
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-methylbenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.