missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-methylimidazole, 95%

Product Code. 15449127
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449127 5 g 5g
15439127 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449127 Supplier Thermo Scientific Alfa Aesar Supplier No. H52407.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16681-59-7 | C4H5BrN2 | 161.002 g/mol

2-Bromo-1-methylimidazole is used in in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1-methylimidazole is used in in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16681-59-7
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.002
MDL Number MFCD02179525
InChI Key BANOTGHIHYMTDL-UHFFFAOYSA-N
Synonym 2-bromo-1-methyl-1h-imidazole, 1-methyl-2-bromoimidazole, 2-bromo-n-methylimidazole, 1h-imidazole, 2-bromo-1-methyl, zlchem 620, pubchem7594, n-methyl-2-bromoimidazole, acmc-209dv3, 2-bromanyl-1-methyl-imidazole, 1-methyl-2-bromo-1h-imidazole
PubChem CID 2773262
IUPAC Name 2-bromo-1-methylimidazole
SMILES CN1C=CN=C1Br

Specifications

Density 1.649
Boiling Point 172°C
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 161
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-methylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.