missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-1-phenyl-1H-imidazole, Thermo Scientific™

Product Code. 10642695
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10642695 5 g 5g
10225954 1 g 1g
10597864 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10642695 Supplier Thermo Scientific Maybridge Supplier No. CC75910DE
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1034566-05-6
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.07
InChI Key KKPQUNLAAJJUCR-UHFFFAOYSA-N
Synonym 2-bromo-1-phenyl-1h-imidazole, 2-bromo-1-phenylimidazole
PubChem CID 45594301

Specifications

Packaging Amber glass bottle
Quantity 5 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 2-bromo-1-phenyl-1H-imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.