missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2',4'-dimethoxyacetophenone, 98%

Product Code. 10022470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10022470 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10022470 Supplier Thermo Scientific Acros Supplier No. 190010010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60965-26-6 | C10H11BrO3 | 259.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 60965-26-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 101°C to 105°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H11BrO3
Linear Formula (CH3O)2C6H3COCH2Br
MDL Number MFCD00000197
Quantity 1 g
Synonym 2-bromo-1-2,4-dimethoxyphenyl ethanone, 2-bromo-2',4'-dimethoxyacetophenone, 2,4-dimethoxyphenacyl bromide, 2-bromo-1-2',4'-dimethoxyphenyl ethanone, 2-bromo-1-2,4-dimethoxyphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethoxyphenyl, 2-bromo-2',4'-dimethoxyacetopheneone, 1-2,4-dimethoxyphenyl-2-bromoethan-1-one, pubchem13433, acmc-1ba4l
Solubility Information Solubility in water: insoluble
InChI Key PKVBZABQCCQHLD-UHFFFAOYSA-N
SMILES COC1=CC(=C(C=C1)C(=O)CBr)OC
IUPAC Name 2-bromo-1-(2,4-dimethoxyphenyl)ethanone
Molecular Weight (g/mol) 259.09
PubChem CID 98683
Formula Weight 259.09
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Bromo-2', 4'-dimethoxyacetophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-2',4'-dimethoxyacetophenone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.