missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-butene, 98%, mixture of cis and trans

Product Code. 10783391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10783391 100 mL Glass bottle 100mL
10479900 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10783391 Supplier Thermo Scientific Acros Supplier No. 201511000
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13294-71-8 | C4H7Br | 135.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13294-71-8
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00000141
InChI Key UILZQFGKPHAAOU-ONEGZZNKSA-N
Synonym 2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene #
PubChem CID 5364387
IUPAC Name (E)-2-bromobut-2-ene
SMILES C\C=C(/C)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown to Yellow
Density 1.3200g/mL
Boiling Point 82°C to 90°C (740.0 mmHg)
Flash Point <1°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=C(Br)CH3
Refractive Index 1.4590 to 1.462
Quantity 100 mL
Beilstein 01, 205
Specific Gravity 1.32
Solubility Information Solubility in water: immiscible
Formula Weight 135
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-butene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-2-butene, mixture of cis and trans
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.