missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-hydroxyacetophenone, 97%

Product Code. 11369929
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369929 25 g 25g
11359929 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369929 Supplier Thermo Scientific Alfa Aesar Supplier No. B25722.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2491-36-3 | C8H7BrO2 | 215.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-36-3
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD01727570
InChI Key SGPKEYSZPHMVNI-UHFFFAOYSA-N
PubChem CID 200671
IUPAC Name 2-bromo-1-(2-hydroxyphenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)O

Specifications

Melting Point 45°C to 47°C
Quantity 25 g
UN Number UN3263
Synonym 2-bromo-2'-hydroxyacetophenone, 2-bromo-1-2-hydroxyphenyl ethanone, 2-hydroxyphenacyl bromide, ethanone, 2-bromo-1-2-hydroxyphenyl, 2-bromo-1-2-hydroxyphenyl ethan-1-one, acetophenone, 2-bromo-2'-hydroxy-6ci,7ci,8ci, ethanone,2-bromo-1-2-hydroxyphenyl, 2-bromoacetyl phenol, acmc-1cg90, 4-08-00-00329 beilstein handbook reference
Formula Weight 215.05
Percent Purity 97%
Chemical Name or Material 2-Bromo-2'-hydroxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.