missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropionic acid, 98%

Product Code. 10652793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10652793 5 g Glass bottle 5g
10185750 100 g Glass bottle 100g
10464555 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10652793 Supplier Thermo Scientific Acros Supplier No. 305200050
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2052-01-9 | C4H7BrO2 | 167.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-01-9
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.01
MDL Number MFCD00004168
InChI Key XXSPGBOGLXKMDU-UHFFFAOYSA-N
Synonym α-Bromoisobutyric acid
IUPAC Name 2-bromo-2-methylpropanoic acid
SMILES CC(C)(Br)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 41°C to 46°C
Color Colorless to Yellow
Boiling Point 198°C to 200°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC(CH3)2CO2H
Quantity 5 g
Merck Index 15, 1431
Solubility Information Solubility in water: slightly soluble
Formula Weight 167.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-methylpropionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.