missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-nitroacetophenone, 98%

Product Code. 15483897
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483897 5 g 5g
15473897 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483897 Supplier Thermo Scientific Alfa Aesar Supplier No. H30275.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6851-99-6 | C8H6BrNO3 | 244.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6851-99-6
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.044
MDL Number MFCD00010294
InChI Key SGXUUCSRVVSMGK-UHFFFAOYSA-N
Synonym 2-bromo-2'-nitroacetophenone, 2'-nitrophenacyl bromide, 2-bromo-1-2-nitrophenyl ethanone, 2-nitrophenacyl bromide, 2-bromo-1-2-nitro-phenyl-ethanone, o-nitrophenacyl bromide, o-nitrophenethyl bromide, 2-o-nitrophenyl ethyl bromide, 2-bromo-1-2-nitrophenyl ethan-1-one, ethanone, 2-bromo-1-2-nitrophenyl
PubChem CID 244025
IUPAC Name 2-bromo-1-(2-nitrophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)[N+](=O)[O-]

Specifications

Melting Point 53°C to 56°C
Odor Pungent
Quantity 5 g
UN Number UN3261
Formula Weight 244.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-nitroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.