missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2'-nitroacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 10776285 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10776285 5 g Glass bottle 5g
10356634 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10776285 Supplier Thermo Scientific Acros Supplier No. 398850050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6851-99-6
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.4
MDL Number MFCD00010294
InChI Key SGXUUCSRVVSMGK-UHFFFAOYSA-N
Synonym 2-bromo-2'-nitroacetophenone, 2'-nitrophenacyl bromide, 2-bromo-1-2-nitrophenyl ethanone, 2-nitrophenacyl bromide, 2-bromo-1-2-nitro-phenyl-ethanone, o-nitrophenacyl bromide, o-nitrophenethyl bromide, 2-o-nitrophenyl ethyl bromide, 2-bromo-1-2-nitrophenyl ethan-1-one, ethanone, 2-bromo-1-2-nitrophenyl
PubChem CID 244025
IUPAC Name 2-bromo-1-(2-nitrophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55.0°C to 57.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4COCH2Br
Quantity 5 g
Formula Weight 244.4
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2'-nitroacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-2'-nitroacetophenone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.