missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-nitropropane-1,3-diol, 98+%

Product Code. 11484743
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484743 500 g 500g
11474743 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484743 Supplier Thermo Scientific Alfa Aesar Supplier No. A11639.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52-51-7 | C3H6BrNO4 | 199.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.

Solubility
Soluble in water(280g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-51-7
Molecular Formula C3H6BrNO4
Molecular Weight (g/mol) 199.988
MDL Number MFCD00007390
InChI Key LVDKZNITIUWNER-UHFFFAOYSA-N
Synonym bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol
PubChem CID 2450
ChEBI CHEBI:31306
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
SMILES C(C(CO)([N+](=O)[O-])Br)O

Specifications

Melting Point ∼122°C to 132°C
pH 5 to 7
Quantity 500 g
UN Number UN3241
Beilstein 1705868
Merck Index 14,1447
Solubility Information Soluble in water(280g/L).
Formula Weight 199.99
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-2-nitropropane-1,3-diol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-2-nitro-1,3-propanediol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Bromo-2-nitro-1,3-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.