missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-hydroxyacetophenone, 96%

Product Code. 10301670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10301670 5 g Glass bottle 5g
10169969 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10301670 Supplier Thermo Scientific Acros Supplier No. 428620050
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2491-37-4 | C8H7BrO2 | 215.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-37-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.04
MDL Number MFCD05664326
InChI Key IEPSGFQQGKPTPM-UHFFFAOYSA-N
PubChem CID 6917206
IUPAC Name 2-bromo-1-(3-hydroxyphenyl)ethanone
SMILES C1=CC(=CC(=C1)O)C(=O)CBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 70°C to 74°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)C6H4COCH2Br
Quantity 5 g
Synonym 2-bromo-1-3-hydroxyphenyl ethanone, 2-bromo-3'-hydroxyacetophenone, 2-bromo-3-hydroxyacetophenone, ethanone, 2-bromo-1-3-hydroxyphenyl, 3-hydroxyphenacyl bromide, 2-bromo-1-3-hydroxyphenyl ethan-1-one, ethanone,2-bromo-1-3-hydroxyphenyl, 2-bromo-3`-hydroxyacetophenone, 2-bromo-3'-hydroxy-acetophenone, tos-bb-1139
Formula Weight 215.04
Percent Purity 96%
Chemical Name or Material 2-Bromo-3′-hydroxyacetophenone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-3'-hydroxyacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.