missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methoxypyridine, 97%, Thermo Scientific™

Product Code. 10546260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
10g; Glass bottle
Product Code.
10546260 10g; Glass bottle
1 options
This item is not returnable. View return policy

Product Code. 10546260

Brand: Thermo Scientific Acros 438150100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24100-18-3
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C6H6BrNO
MDL Number MFCD01570896
Quantity 10g
Synonym 2-bromo-3-methoxy-pyridine, pyridine, 2-bromo-3-methoxy, 2-bromo-3-methoxy pyridine, pubchem5916, acmc-1cefu, ksc552s2b, #, pyridine,2-bromo-3-methoxy, 2-bromo-3-methoxypyridine, abbypharma ap-17-5948
InChI Key PDOWLYNSFYZIQX-UHFFFAOYSA-N
SMILES COC1=C(N=CC=C1)Br
IUPAC Name 2-bromo-3-methoxypyridine
Molecular Weight (g/mol) 188.02
PubChem CID 90364
Formula Weight 188.02
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-methoxypyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-3-methoxypyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.