missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-3-methylpyridine-5-boronic acid, 97%, Thermo Scientific™

Product Code. 10021244
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10021244 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10021244 Supplier Thermo Scientific Maybridge Supplier No. MAY00055DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1003043-34-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H7BBrNO2
MDL Number MFCD08274468
Quantity 1 g
Synonym 2-bromo-3-methylpyridine-5-boronic acid, 6-bromo-5-methylpyridine-3-boronic acid, 6-bromo-5-methylpyridin-3-yl boronic acid, 2-bromo-3-methyl-5-pyridineboronic acid, 5-borono-2-bromo-3-picoline, 6-bromo-5-methylpyridin-3-yl-3-boronic acid, 2-bromo-3-picoline-5-boronic acid, 6-bromo-5-methyl-3-pyridyl boronic acid, pubchem5081, acmc-2097oc
InChI Key IPCXBKSYMWZLBR-UHFFFAOYSA-N
SMILES B(C1=CC(=C(N=C1)Br)C)(O)O
IUPAC Name (6-bromo-5-methylpyridin-3-yl)boronic acid
Molecular Weight (g/mol) 215.841
PubChem CID 17750219
Formula Weight 215.84
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-methylpyridine-5-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.