missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylthiophene, 97%

Product Code. 11397746
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397746 25 g 25g
11387746 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397746 Supplier Thermo Scientific Alfa Aesar Supplier No. L11393.14
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14282-76-9 | C5H5BrS | 177.059 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14282-76-9
Molecular Formula C5H5BrS
Molecular Weight (g/mol) 177.059
MDL Number MFCD00059741
InChI Key YYJBWYBULYUKMR-UHFFFAOYSA-N
Synonym thiophene, 2-bromo-3-methyl, 2-bromo-3-methyl-thiophene, 1-bromo-2-methyl thiophene, pubchem5191, 2-bromo-3methylthiophene, 2-bromo-3-methythiophene, acmc-1buh4, 2-bromo-3methyl thiophene, 2-bromo-3-methyl thiophene, ksc491i3r
PubChem CID 84314
IUPAC Name 2-bromo-3-methylthiophene
SMILES CC1=C(SC=C1)Br

Specifications

Density 1.58
Boiling Point 173°C to 176°C
Flash Point 68°C (154°F)
Refractive Index 1.571
Quantity 25 g
UN Number UN2810
Beilstein 105719
Formula Weight 177.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-methylthiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.