missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromo-3-thienyl)methylamine, 97%, Thermo Scientific™

Product Code. 10611703
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10611703 1 g Amber Glass Bottle 1g
10315112 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10611703 Supplier Thermo Scientific Maybridge Supplier No. SPB08448DA
 Restricted Item
Estimated Shipment: 08-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 157664-47-6
Molecular Formula C5H6BrNS
Molecular Weight (g/mol) 192.074
MDL Number MFCD04971979
InChI Key SLKZUJBSIKGHFU-UHFFFAOYSA-N
Synonym 2-bromo-3-thienyl methylamine, 2-bromothiophen-3-yl methanamine, 2-bromo-3-thiophenemethylamine, 3-thiophenemethanamine,2-bromo, 1-2-bromothiophen-3-yl methanamine, 2-bromothien-3-yl methylamine hydrochloride, pubchem5276, h-val-ssna, bestipharma 583-851, 3-aminomethyl-2-bromothiophene
PubChem CID 2735586
IUPAC Name (2-bromothiophen-3-yl)methanamine
SMILES C1=CSC(=C1CN)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 228.54
Percent Purity 97%
Chemical Name or Material (2-Bromo-3-thienyl)methylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.