missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-thiophenecarboxylic acid, 96%

Product Code. 10336304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10336304 1 g Glass bottle 1g
10051554 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336304 Supplier Thermo Scientific Acros Supplier No. 426840010
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24287-95-4 | C5H3BrO2S | 207.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24287-95-4
Molecular Formula C5H3BrO2S
Molecular Weight (g/mol) 207.04
MDL Number MFCD05865213
InChI Key RVSXMPCELBYUSF-UHFFFAOYSA-N
Synonym 2-bromo-3-thiophenecarboxylic acid, 2-bromo-thiophene-3-carboxylic acid, 2-bromo-thiophene-3-carboxylicacid, 3-thiophenecarboxylic acid, 2-bromo, 2-bromo-3-thenoic acid, acmc-209xc0, 2-bromo-3-carboxythiophene, 2-bromothiophene-3-carboxylic?acid, 2-bromo-3-thiophene carboxylic acid, 2-bromo-thiophene 3-carboxylic acid
PubChem CID 12320971
IUPAC Name 2-bromothiophene-3-carboxylic acid
SMILES C1=CSC(=C1C(=O)O)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 179.0°C to 181.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 207.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-thiophenecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-3-thiophenecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.