missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-chlorophenol, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-4-chlorophenol |
| CAS | 695-96-5 |
| Molecular Formula | C6 H4 Br Cl O |
| Quantity | 10 g |
| Synonym | Phenol, 2-bromo-4-chloro-, 2-Bromo-4-chlorophenol, NSC 59695 |
| InChI Formula | InChI=1S/C6H4BrClO/c7-5-3-4(8)1-2-6(5)9/h1-3,9H |
| SMILES | Oc1ccc(Cl)cc1Br |
| IUPAC Name | 2-bromo-4-chlorophenol |
| Molecular Weight (g/mol) | 207.45 |
| Formula Weight | 205.9134 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction