missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-(diethylamino)acetophenone, 98%

Product Code. 11318056
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318056 5 g 5g
11308056 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318056 Supplier Thermo Scientific Alfa Aesar Supplier No. L13337.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 207986-25-2 | C12H16BrNO | 270.17 g/mol

2-Bromo-4'-(diethylamino)acetophenone, is used as a pharmaceutical intermediate. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-(diethylamino)acetophenone, is used as a pharmaceutical intermediate. It is also used as a primary and secondary intermediate.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207986-25-2
Molecular Formula C12H16BrNO
Molecular Weight (g/mol) 270.17
MDL Number MFCD00151798
InChI Key ALTMCDJIFXTEPV-UHFFFAOYSA-N
Synonym 2-bromo-1-4-diethylamino phenyl ethanone, alpha-bromo-4-diethylamino acetophenone, 2-bromo-4'-diethylamino acetophenone, 2-bromo-1-4-diethylamino phenyl ethan-1-one, 4-diethylaminophenacyl bromide, n,n-diethyl-4-bromoacetyl aniline, 4-n,n-diethylamino phenacyl bromide, .alpha.-bromo-4-diethylaminoacetophenone, 2-bromo-1-4-diethylaminophenyl ethanone
PubChem CID 2795030
IUPAC Name 2-bromo-1-[4-(diethylamino)phenyl]ethanone
SMILES CCN(CC)C1=CC=C(C=C1)C(=O)CBr

Specifications

Melting Point 50°C to 52°C
Quantity 5 g
Beilstein 7921481
Solubility Information It is insoluble in water.
Formula Weight 270.18
Percent Purity 98%
Chemical Name or Material 2-Bromo-4'-(diethylamino)acetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.