missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-fluoroacetophenone, 97%

Product Code. 11488206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488206 25 g 25g
11304729 1 g 1g
11478206 5 g 5g
11498206 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11488206 Supplier Thermo Scientific Alfa Aesar Supplier No. A15319.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 403-29-2 | C8H6BrFO | 217.037 g/mol

2-Bromo-4'-fluoroacetophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4′-fluoroacetophenone is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-29-2
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
MDL Number MFCD00040830
InChI Key ZJFWCELATJMDNO-UHFFFAOYSA-N
Synonym 2-bromo-4'-fluoroacetophenone, 4-fluorophenacyl bromide, 2-bromo-1-4-fluorophenyl ethanone, 2-bromo-1-4-fluorophenyl ethan-1-one, p-fluorophenacyl bromide, 2'-bromo-4-fluoroacetophenone, ethanone, 2-bromo-1-4-fluorophenyl, bromo-4-fluoroacetophenone, 4-fluorophenacylbromide, 2-bromo-1-4-fluoro-phenyl-ethanone
PubChem CID 96749
IUPAC Name 2-bromo-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)F

Specifications

Melting Point 45°C to 49°C
Boiling Point 150°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 637863
Solubility Information Soluble in methanol.
Formula Weight 217.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-4'-fluoroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.