missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-hydroxyacetophenone, TRC

Product Code. 30342638
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342638 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342638 Supplier Toronto Research Chemicals Supplier No. TRCB68445010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Bromo-4'-hydroxyacetophenone
CAS 2491-38-5
Molecular Formula C8 H7 Br O2
Quantity 10 g
Synonym Ethanone, 2-bromo-1-(4-hydroxyphenyl)-, Acetophenone, 2-bromo-4'-hydroxy- (6CI,7CI,8CI), 2-Bromo-1-(4-hydroxyphenyl)ethanone, 1-(4-Hydroxyphenyl)-2-bromoethanone, 2-Bromo-1-(4-hydroxyphenyl)ethan-1-one, 2-Bromo-4'-hydroxyacetophenone, 4-Hydroxyphenacyl bromide, 4'-Hydroxy-2-bromoacetophenone, Busan 1130, Busan 90, Butrol 1130, p-Hydroxyphenacyl bromide, α-Bromo-4'-hydroxyacetophenone, α-Bromo-p-hydroxyacetophenone, ω-Bromo-p-hydroxyacetophenone
InChI Formula InChI=1S/C8H7BrO2/c9-5-8(11)6-1-3-7(10)4-2-6/h1-4,10H,5H2
SMILES Oc1ccc(cc1)C(=O)CBr
IUPAC Name 2-bromo-1-(4-hydroxyphenyl)ethanone
Molecular Weight (g/mol) 215.04
Formula Weight 213.9629
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.