missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methyl-1,3-thiazole-5-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10446472
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10446472

Brand: Thermo Scientific Maybridge MO00751DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 40003-41-6
Molecular Formula C5H4BrNO2S
Molecular Weight (g/mol) 222.056
MDL Number MFCD03822128
InChI Key HMSQZHBSTZZNGI-UHFFFAOYSA-N
Synonym 2-bromo-4-methylthiazole-5-carboxylic acid, 2-bromo-4-methyl-thiazole-5-carboxylic acid, 2-bromo-4-methylthiazol-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-bromo-4-methyl, maybridge3_004237, pubchem23205, acmc-209x4s, 2-bromo-4-methyl-5-thiazolecarboxylic acid, 2-bromo-4-methyl-5-thiazole carboxylic acid
PubChem CID 2824058
IUPAC Name 2-bromo-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)Br)C(=O)O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 222.06
Percent Purity 90%
Chemical Name or Material 2-Bromo-4-methyl-1, 3-thiazole-5-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.