missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methyl-1,3-thiazole-5-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10533052
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10533052 10 g Amber Glass Bottle 10g
10446472 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533052 Supplier Thermo Scientific Maybridge Supplier No. MO00751EA
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 40003-41-6
Molecular Formula C5H4BrNO2S
Molecular Weight (g/mol) 222.056
MDL Number MFCD03822128
InChI Key HMSQZHBSTZZNGI-UHFFFAOYSA-N
Synonym 2-bromo-4-methylthiazole-5-carboxylic acid, 2-bromo-4-methyl-thiazole-5-carboxylic acid, 2-bromo-4-methylthiazol-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-bromo-4-methyl, maybridge3_004237, pubchem23205, acmc-209x4s, 2-bromo-4-methyl-5-thiazolecarboxylic acid, 2-bromo-4-methyl-5-thiazole carboxylic acid
PubChem CID 2824058
IUPAC Name 2-bromo-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)Br)C(=O)O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 222.06
Percent Purity 90%
Chemical Name or Material 2-Bromo-4-methyl-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.