missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4'-methylpropiophenone, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-4'-methylpropiophenone |
| CAS | 1451-82-7 |
| Molecular Formula | C10 H11 Br O |
| Quantity | 250 mg |
| Synonym | 2-Bromo-1-(4-methylphenyl)-1-propanone, 4-Methylphenyl 1-Bromoethyl Ketone |
| InChI Formula | InChI=1S/C10H11BrO/c1-7-3-5-9(6-4-7)10(12)8(2)11/h3-6,8H,1-2H3 |
| SMILES | CC(Br)C(=O)c1ccc(C)cc1 |
| IUPAC Name | 2-bromo-1-(4-methylphenyl)propan-1-one |
| Molecular Weight (g/mol) | 227.1 |
| Formula Weight | 225.9993 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction