missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylpyrimidine, 97%

Product Code. 15450788
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450788 25 g 25g
15430788 1 g 1g
15440788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450788 Supplier Thermo Scientific Alfa Aesar Supplier No. H54503.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 130645-48-6 | C5H5BrN2 | 173.013 g/mol

2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 130645-48-6
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD04037883
InChI Key UZIJEBOLOXOVFY-UHFFFAOYSA-N
PubChem CID 10942928
IUPAC Name 2-bromo-4-methylpyrimidine
SMILES CC1=NC(=NC=C1)Br

Tekniske data

Melting Point 45°C to 50°C
Quantity 25 g
Synonym 2-bromo-4-methyl-pyrimidine, pyrimidine, 2-bromo-4-methyl, pyrimidine,2-bromo-4-methyl, pubchem7007, acmc-209z1w, 2-bromo-4-methlypyrimidine, 2-bromo-4-methyl pyrimidine, 2-bromo-4-methylpyrimidine
Solubility Information Slightly soluble in water.
Formula Weight 173.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-methylpyrimidine

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.