missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-(trifluoromethyl)aniline, 98%, BLD Pharm™

Product Code. 31111533
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31111533 10 g
31111231 25 g
31101884 100 g
31110466 5 g
31100665 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31111533 Supplier BLD Pharm Supplier No. BD1032810g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 57946-63-1
Molecular Formula C7H5BrF3N
MDL Number MFCD00042150
Quantity 10 g
SMILES NC1=CC=C(C(F)(F)F)C=C1Br
Molecular Weight (g/mol) 240.02
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.